BKM120 ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(5-[2,6-Di(4-morpholinyl)-4-pyrimidinyl]-4-(trifluoromethyl)-2-pyridinamine, 5-(2,6-Dimorpholinopyrimidin-4-yl)-4-(trifluoromethyl)pyridin-2-amine, Buparlisib, NVP-BKM120
A105286-10MG
A105286-1MG
A105286-5MG
A105286-50MG
76821-522EA
40.32
USD
76821-522
76821-524
76821-526
77682-969
BKM120 ≥98%
BKM120
Precaución: For research use only.
Formula:
C₁₈H₂₁F₃N₆O₂ MW: 410.4 g/mol |
MDL Number:
MFCD18251596 CAS Number: 944396-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...