(6-Methoxypyridin-3-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A104912-1G A104912-250MG A104912-5G
76821-500EA 50 USD
76821-500 76821-502 76821-504
(6-Methoxypyridin-3-yl)methanamine dihydrochloride 95%
(6-Methoxypyridin-3-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂O
MDL Number: MFCD23135657
CAS Number: 169045-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR