3,5-Diiodo-L-tyrosine dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr(3,5-DiI)-OH·2H₂O, (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate

A104901-10G A104901-5G
76821-496EA 9.55 USD
76821-496 76821-498
3,5-Diiodo-L-tyrosine dihydrate 98%
3,5-Diiodo-L-tyrosine dihydrate

Precaución: for Research Use Only

Formula: C₉H₉I₂NO₃·2H₂O
MW: 469.01 g/mol
MDL Number: MFCD00016542
CAS Number: 18835-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR