1-Ethyl-2,3,3-trimethyl-3H-indol-1-ium-5-sulfonate 95%

Supplier: Ambeed
Warning

A104711-100MG A104711-250MG
76821-484EA 39 USD
76821-484 76821-486
1-Ethyl-2,3,3-trimethyl-3H-indol-1-ium-5-sulfonate 95%
1-Ethyl-2,3,3-trimethyl-3H-indol-1-ium-5-sulfonate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃S
MDL Number: MFCD07369396
CAS Number: 146368-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR