Methyl 2,6-dichloro-5-methoxypyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A104583-100MG A104583-1G A104583-250MG A104583-5G
76821-446EA 53.05 USD
76821-446 76821-448 76821-450 76821-452
Methyl 2,6-dichloro-5-methoxypyrimidine-4-carboxylate 98%
Methyl 2,6-dichloro-5-methoxypyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₃
MW: 237.04 g/mol
MDL Number: MFCD16660462
CAS Number: 878650-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR