3-Fluoro-4-(trifluoromethoxy)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Fluoro-4-(trifluoromethoxy)phenyl)boronic acid

A104488-1G A104488-250MG A104488-10G A104488-5G
76821-440EA 67.9 USD
76821-440 76821-442 77682-959 77682-960
3-Fluoro-4-(trifluoromethoxy)phenylboronic acid 97%
3-Fluoro-4-(trifluoromethoxy)phenylboronic acid

Precaución: For research use only.

Formula: C₇H₅BF₄O₃
MW: 223.920 g/mol
MDL Number: MFCD07368680
CAS Number: 187804-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR