2,4-Dichloro-3-cyano-5-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A104179-1G A104179-5G
76821-422EA 88.06 USD
76821-422 76821-424
2,4-Dichloro-3-cyano-5-fluorobenzoic acid 98%
2,4-Dichloro-3-cyano-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₂Cl₂FNO₂
MW: 234.01 g/mol
MDL Number: MFCD18801150
CAS Number: 117528-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR