Methyl 5-bromo-6-methoxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: 5-bromo-6-methoxy-3-pyridinecarboxylic acid methyl ester

A104068-100MG A104068-10G A104068-1G A104068-250MG A104068-25G A104068-5G
76821-408EA 6 USD
76821-408 76821-410 76821-412 76821-414 76821-416 76821-418
Methyl 5-bromo-6-methoxynicotinate 98%
Methyl 5-bromo-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MDL Number: MFCD14698128
CAS Number: 93349-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR