2-Aminopyridine-6-methanol 97%

Supplier: Ambeed
Warning

Synonyms: (6-Aminopyridin-2-yl)methanol

A104042-10G A104042-1G A104042-5G
76821-402EA 293.87 USD
76821-402 76821-404 76821-406
2-Aminopyridine-6-methanol 97%
2-Aminopyridine-6-methanol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD09863097
CAS Number: 79651-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR