(S)-1-(4-Methoxyphenyl)ethanol 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A103967-1G A103967-5G A103967-250MG
76821-398EA 35 USD
76821-398 77565-804 77682-953
(S)-1-(4-Methoxyphenyl)ethanol 97%, Ambeed.Inc
(S)-1-(4-Methoxyphenyl)ethanol

Precaución: For research use only.

Formula: C₉H₁₂O₂
MDL Number: MFCD09863666
CAS Number: 1572-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR