6-Hydroxy-5-iodopyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A103909-100MG A103909-10G A103909-1G A103909-250MG A103909-25G A103909-5G
76821-382EA 11.67 USD
76821-382 76821-384 76821-386 76821-388 76821-390 76821-392
6-Hydroxy-5-iodopyrimidin-4(3H)-one 95%
6-Hydroxy-5-iodopyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₄H₃IN₂O₂
MW: 237.9833 g/mol
MDL Number: MFCD21742320
CAS Number: 1429056-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR