3,4-Dimethylcyclopent-2-enone 95%

Supplier: Ambeed
Danger

A103831-250MG A103831-5G
76821-370EA 74 USD
76821-370 76821-372
3,4-Dimethylcyclopent-2-enone 95%
3,4-Dimethylcyclopent-2-enone

Precaución: for Research Use Only

Formula: C₇H₁₀O
MDL Number: MFCD09701445
CAS Number: 30434-64-1
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR