4-Bromo-1-cyclopropyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

A103701-100MG A103701-1G A103701-250MG
76821-362EA 23.34 USD
76821-362 76821-364 76821-366
4-Bromo-1-cyclopropyl-1H-pyrazole 95%
4-Bromo-1-cyclopropyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
MDL Number: MFCD18250639
CAS Number: 1151802-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR