Diethyl 5-bromoisophthalate 97%

Supplier: Ambeed
Warning

A103593-1G A103593-250MG A103593-25G A103593-5G
76821-354EA 18.04 USD
76821-354 76821-356 76821-358 76821-360
Diethyl 5-bromoisophthalate 97%
Diethyl 5-bromoisophthalate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrO₄
MW: 301.13 g/mol
MDL Number: MFCD12913556
CAS Number: 127437-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR