2-(Trimethylsilylethynyl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: (2-Pyridylethynyl)trimethylsilane, 2-[(Trimethylsilyl)ethynyl]pyridine, 2-((Trimethylsilyl)ethynyl)pyridine

A103592-1G A103592-250MG A103592-25G A103592-5G
76821-346EA 11 USD
76821-346 76821-348 76821-350 76821-352
2-(Trimethylsilylethynyl)pyridine 95%
2-(Trimethylsilylethynyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NSi
MDL Number: MFCD00066349
CAS Number: 86521-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR