4-Amino-2,3,5-trichloropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2,3,5-trichloropyridin-4-amine

A103374-1G A103374-250MG
76821-334EA 438.16 USD
76821-334 76821-336
4-Amino-2,3,5-trichloropyridine 95%
4-Amino-2,3,5-trichloropyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₃N₂
MW: 197.45 g/mol
MDL Number: MFCD00153908
CAS Number: 28443-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR