Pyridoxal-5'-phosphate 98%
Supplier: Ambeed
|
Warning
|
A103025-100G
A103025-10G
A103025-1G
A103025-25G
A103025-5G
A103025-500G
76821-324EA
103
USD
76821-324
76821-326
76821-328
76821-330
76821-332
77884-933
Pyridoxal-5'-phosphate 98%
Pyridoxal 5'-phosphate
Precaución: for Research Use Only
Formula:
C₈H₁₀NO₆P |
MDL Number:
MFCD00006333 CAS Number: 54-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...