Methyl-1,3-benzoxazole-7-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl benzo[d]oxazole-7-carboxylate

A102984-100MG A102984-250MG
76821-320EA 15.92 USD
76821-320 76821-322
Methyl-1,3-benzoxazole-7-carboxylate 95%
Methyl-1,3-benzoxazole-7-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.16 g/mol
MDL Number: MFCD11042246
CAS Number: 1086378-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR