Ethyl 5-chlorothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A102812-1G A102812-250MG A102812-5G
76821-304EA 36.08 USD
76821-304 76821-306 76821-308
Ethyl 5-chlorothiazole-4-carboxylate 97%
Ethyl 5-chlorothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD22494956
CAS Number: 425392-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR