3-(2-Ethylhexyl)thiophene 97% (by GC)

Supplier: Ambeed
Warning

A102793-1G A102793-250MG A102793-25G
76821-298EA 22.28 USD
76821-298 76821-300 76821-302
3-(2-Ethylhexyl)thiophene 97% (by GC)
3-(2-Ethylhexyl)thiophene

Precaución: for Research Use Only

Formula: C₁₂H₂₀S
MW: 196.35 g/mol
MDL Number: MFCD02260336
CAS Number: 121134-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR