4-Nitrophenyl (thiazol-5-ylmethyl) carbonate hydrochloride 95%

Supplier: Ambeed
Warning

A102678-5G A102678-1G A102678-25G
76821-292EA 39 USD
76821-292 77884-929 77884-930
4-Nitrophenyl (thiazol-5-ylmethyl) carbonate hydrochloride 95%
4-Nitrophenyl (thiazol-5-ylmethyl) carbonate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₉ClN₂O₅S
MDL Number: MFCD09832876
CAS Number: 154212-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR