4-Hydroxy-2-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A102261-1G A102261-250MG A102261-5G
76821-280EA 210.06 USD
76821-280 76821-282 77548-930
4-Hydroxy-2-iodobenzaldehyde 97%
4-Hydroxy-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅IO₂
MW: 248.01 g/mol
MDL Number: MFCD16999762
CAS Number: 90151-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR