(2-Formyl-5-(trifluoromethyl)phenyl)boronic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Formyl-5-(trifluoromethyl)phenylboronic acid
A994210-100MG
A994210-1G
A994210-250MG
76821-170EA
18
USD
76821-170
76821-172
77884-913
(2-Formyl-5-(trifluoromethyl)phenyl)boronic acid ≥95%
(2-Formyl-5-(trifluoromethyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₈H₆BF₃O₃ |
MDL Number:
MFCD11617253 CAS Number: 1204580-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...