4-Benzyloxy-2-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A991503-100G A991503-1G A991503-250MG A991503-25G A991503-5G
76821-148EA 1324 USD
76821-148 76821-150 76821-152 76821-154 76821-156
4-Benzyloxy-2-fluorobenzoic acid 98%
4-Benzyloxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₃
MDL Number: MFCD12407082
CAS Number: 114045-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR