4-(Pyrrolidin-1-ylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Pyrrolidin-1-ylsulphonyl)phenyl)boronic acid

A988612-1G A988612-250MG A988612-5G
76821-140EA 131.56 USD
76821-140 76821-142 76821-144
4-(Pyrrolidin-1-ylsulfonyl)benzeneboronic acid 98%
4-(Pyrrolidin-1-ylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₄S
MW: 255.098 g/mol
MDL Number: MFCD06659847
CAS Number: 486422-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR