2,4-Difluoro-1-isopropoxybenzene 95+%

Supplier: Ambeed
Warning

Synonyms: 2-(2',4'-Difluorophenoxy)propane

A981994-1G A981994-250MG A981994-5G
76821-128EA 124.13 USD
76821-128 76821-130 76821-132
2,4-Difluoro-1-isopropoxybenzene 95+%
2,4-Difluoro-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
MW: 172.17 g/mol
MDL Number: MFCD00070792
CAS Number: 203059-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR