4-Amino-2-methoxy-5-nitropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-5-nitropyridin-4-amine

A981561-100MG A981561-1G A981561-250MG
76821-126EA 64.72 USD
76821-126 76982-056 76982-058
4-Amino-2-methoxy-5-nitropyridine 95%
4-Amino-2-methoxy-5-nitropyridine

Precaución: For research use only

Formula: C₆H₇N₃O₃
MW: 169.138 g/mol
MDL Number: MFCD11977410
CAS Number: 127356-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR