4-Chloro-6-methylpyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-6-methylpicolinic acid

A978179-100MG A978179-250MG
76821-108EA 108.22 USD
76821-108 76821-110
4-Chloro-6-methylpyridine-2-carboxylic acid 98%
4-Chloro-6-methylpyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD07374951
CAS Number: 30235-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR