5-Bromo-2-thiophenesulphonyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-thiophenesulphonyl Chloride, 5-Bromothiophene-2-sulphonyl chloride

A975901-1G A975901-25G A975901-5G
76821-102EA 9 USD
76821-102 76821-104 76821-106
5-Bromo-2-thiophenesulphonyl chloride 96%
5-Bromo-2-thiophenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₄H₂BrClO₂S₂
MW: 261.55 g/mol
MDL Number: MFCD00084909
CAS Number: 55854-46-1
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR