1,2,3,4-Tetrahydroquinoline-2-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A967279-100MG A967279-1G A967279-250MG
76821-090EA 38.2 USD
76821-090 76821-092 76821-094
1,2,3,4-Tetrahydroquinoline-2-carboxylic acid hydrochloride 95%
1,2,3,4-Tetrahydroquinoline-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
MW: 213.66 g/mol
MDL Number: MFCD09878848
CAS Number: 75493-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR