Methyl-2-amino-6-chloronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-6-chloronicotinate

A966937-100MG A966937-250MG A966937-1G A966937-5G
76821-086EA 22.28 USD
76821-086 76821-088 77614-684 77674-422
Methyl-2-amino-6-chloronicotinate 97%
Methyl-2-amino-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.6 g/mol
MDL Number: MFCD17015967
CAS Number: 849805-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR