2-Methyl-1H-benzoimidazole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A963174-1G A963174-250MG A963174-5G A963174-25G
76821-080EA 9 USD
76821-080 76821-082 76821-084 77884-901
2-Methyl-1H-benzoimidazole-5-carboxylic acid 98%
2-Methyl-1H-benzoimidazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD00800488
CAS Number: 709-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR