4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-3-(isopropylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(isopropylcarbamoyl)benzeneboronic acid, 4-Fluoro-3-(isopropylcarbamoyl)phenylboronic acid

A957894-1G A957894-250MG A957894-5G
76821-060EA 43.5 USD
76821-060 76821-062 76821-064
4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid 98%
4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BFNO₃
MW: 225.03 g/mol
MDL Number: MFCD08235040
CAS Number: 874219-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR