2,4,6-Trichloronicotinonitrile 95%

Supplier: Ambeed
Warning

A957402-250MG A957402-1G A957402-100MG A957402-5G
76821-046EA 95 USD
76821-046 77595-544 77884-894 77884-895
2,4,6-Trichloronicotinonitrile 95%
2,4,6-Trichloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆HCl₃N₂
MDL Number: MFCD19707185
CAS Number: 40381-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR