2-Methoxycinnamaldehyde 98%

Supplier: Ambeed
Warning

A949113-100G A949113-10G A949113-25G A949113-500G
76821-030EA 53.05 USD
76821-030 76821-032 76821-034 77674-412
2-Methoxycinnamaldehyde 98%
2-Methoxycinnamaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.1852 g/mol
MDL Number: MFCD00007001
CAS Number: 1504-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR