(3-Bromo-5-fluoro-2-methoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-fluoro-2-methoxyphenylboronic acid

A945086-1G A945086-250MG A945086-5G
76821-024EA 57 USD
76821-024 76821-026 76821-028
(3-Bromo-5-fluoro-2-methoxyphenyl)boronic acid 95%
(3-Bromo-5-fluoro-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₃
MDL Number: MFCD06411347
CAS Number: 352525-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR