4-Bromo-5-methyl-2-nitroaniline 95%

Supplier: Ambeed
Warning

A944346-1G A944346-250MG A944346-5G
76821-016EA 32.89 USD
76821-016 76821-018 76821-020
4-Bromo-5-methyl-2-nitroaniline 95%
4-Bromo-5-methyl-2-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD16619131
CAS Number: 827-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR