4-Chloro-1H-pyrrolo[2,3-c]pyridine 95%

Supplier: Ambeed
Warning

A943954-250MG A943954-1G
76821-006EA 27.59 USD
76821-006 77595-530
4-Chloro-1H-pyrrolo[2,3-c]pyridine 95%
4-Chloro-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MDL Number: MFCD16657181
CAS Number: 1188313-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR