3-(Boc-amino)-2,2-dimethylpropionic Acid 98%

Supplier: Ambeed
Warning

A939994-100MG A939994-1G A939994-250MG A939994-5G
76820-998EA 12 USD
76820-998 76821-000 76821-002 76821-004
3-(Boc-amino)-2,2-dimethylpropionic Acid 98%
3-(Boc-amino)-2,2-dimethylpropionic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD09952625
CAS Number: 180181-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR