4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one 96%

Supplier: Ambeed
Warning

A929742-10G A929742-25G A929742-5G
76820-984EA 33.95 USD
76820-984 76820-986 76820-988
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one 96%
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one

Precaución: for Research Use Only

Formula: C₅H₆O₄
MW: 130.099 g/mol
MDL Number: MFCD12165896
CAS Number: 91526-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR