Cyclohexylmethyl cyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

A928297-10G A928297-1G A928297-5G
76820-976EA 353.28 USD
76820-976 76820-978 76820-980
Cyclohexylmethyl cyclohexanecarboxylate 95%
Cyclohexylmethyl cyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₄O₂
MW: 224.34 g/mol
MDL Number: MFCD20528370
CAS Number: 2611-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR