4-Chlorobutan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A926336-100MG A926336-250MG A926336-1G A926336-5G
76820-972EA 39 USD
76820-972 76820-974 77614-646 77674-382
4-Chlorobutan-1-amine hydrochloride 95%
4-Chlorobutan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁Cl₂N
MDL Number: MFCD07780199
CAS Number: 6435-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR