Triphenylphosphine copper(I) hydride hexamer 95%

Supplier: Ambeed
Danger

Synonyms: Stryker's reagent

A917462-1G A917462-250MG A917462-5G
76820-962EA 162.32 USD
76820-962 76820-964 77595-476
Triphenylphosphine copper(I) hydride hexamer 95%
Triphenylphosphine copper(I) hydride hexamer

Precaución: for Research Use Only

Formula: C₁₀₈H₁₀₂Cu₆P₆
MW: 1967.08 g/mol
MDL Number: MFCD00221518
CAS Number: 33636-93-0
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR