N1-Methylbenzene-1,4-diamine 95%

Supplier: Ambeed
Warning

A911850-1G A911850-5G A911850-250MG
76820-950EA 130 USD
76820-950 76820-952 77674-350
N1-Methylbenzene-1,4-diamine 95%
N1-Methylbenzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00044895
CAS Number: 623-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR