5-Chloro-2-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

A909792-100G A909792-1G A909792-250MG A909792-5G A909792-25G
76820-932EA 270 USD
76820-932 76820-934 76820-936 76820-938 77595-468
5-Chloro-2-methyl-3-nitropyridine 98%
5-Chloro-2-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD18255264
CAS Number: 1211533-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR