6-Bromo-3-chloro-2-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A906050-100MG A906050-1G
76820-920EA 40.32 USD
76820-920 76820-922
6-Bromo-3-chloro-2-fluorobenzaldehyde 98%
6-Bromo-3-chloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
MW: 237.454 g/mol
MDL Number: MFCD11520661
CAS Number: 1114809-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR