2-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanenitrile 98%

Supplier: Ambeed
Warning

A905781-100MG A905781-250MG
76820-916EA 125.19 USD
76820-916 76820-918
2-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanenitrile 98%
2-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanenitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₂BNO₂
MW: 271.16 g/mol
MDL Number: MFCD19689513
CAS Number: 1082066-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR