4-Carboxy-3-nitrophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Borono-2-nitrobenzoic acid, 3-Nitro-4-carboxyphenylboronic acid, 4-Carboxy-3-nitrobenzeneboronic acid

A896529-250MG A896529-1G A896529-5G
76820-908EA 6 USD
76820-908 77674-332 77674-334
4-Carboxy-3-nitrophenylboronic acid 95%
4-Carboxy-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BNO₆
MDL Number: MFCD10696661
CAS Number: 80500-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR