(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-[(1R,2R)-1,2-Diaminoethylene]bisphenol, (1R,2R)-1,2-Diamino-1,2-bis(2-hydroxyphenyl)ethane, 2,2'-((1R,2R)-1,2-Diaminoethane-1,2-diyl)diphenol

A895763-250MG A895763-1G A895763-10G A895763-25G A895763-5G
76820-906EA 57 USD
76820-906 77674-328 77884-841 77884-842 77884-843
(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine 98%
(1R,2R)-1,2-Bis(2-hydroxyphenyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
MDL Number: MFCD09751760
CAS Number: 870991-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR