2,6-Dichloropyrimidine-4-carboxamide 96%

Supplier: Ambeed
Warning

A891011-100MG A891011-1G A891011-250MG A891011-5G
76820-888EA 49.87 USD
76820-888 76820-890 76820-892 76820-894
2,6-Dichloropyrimidine-4-carboxamide 96%
2,6-Dichloropyrimidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N₃O
MW: 192.00 g/mol
MDL Number: MFCD09881209
CAS Number: 98136-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR